Media/Storage Bottles, Amber, United Scientific Supplies
Supplier: United Scientific Supplies
BMA0100 BMA0250 BMA0500 BMA1000
76531-426PK 226.64 USD
76531-426 76531-428 76531-430 76531-432
Media/Storage Bottles, Amber, United Scientific Supplies
Bottles Glass Bottles
Pack of 10 borosilicate glass media / storage bottles.
  • Amber color designed to protect light-sensitive materials during transport and storage
  • Feature printed white graduations (from 40 - 80 ml) and durable marking spots
  • Maximum use temperature is 450 °C (842 °F)
  • Autoclavable
Included GL 45 cap is linerless, both the caps and drip-free rings are made from polypropylene and autoclavable to 140 °C.