Beakers, Low Form, Borosilicate Glass, United Scientific Supplies
Supplier: United Scientific Supplies
BG1000-10 BG1000-100 BG1000-1000 BG1000-150 BG1000-1500 BG1000-20 BG1000-2000 BG1000-25 BG1000-250 BG1000-30 BG1000-400 BG1000-4000 BG1000-5 BG1000-50 BG1000-500 BG1000-5000 BG1000-600 BG1000-800
76531-342PK 30.04 USD
76531-342 76531-344 76531-346 76531-348 76531-350 76531-352 76531-354 76531-356 76531-358 76531-360 76531-362 76531-364 76531-366 76531-368 76531-370 76531-372 76531-374 76531-376
Beakers, Low Form, Borosilicate Glass, United Scientific Supplies
Beakers
Made from low-expansion borosilicate glass, these beakers feature heavy and uniform wall thickness, spouts designed for easy pouring, and beaded tops. The 20ml and larger sizes also have a double graduated metric scale and a marking area.
  • Size ranges from 100 to 5000 ml
  • Thicker walls than traditonal beakers
Tolerance for beakers is approximately +/- 5%.