CHO Host Cell Protein Standards
Supplier: Enzo Life Sciences
PRT122-0025
76529-712EA 706.32 USD
76529-712
CHO Host Cell Protein Standards
Proteins and Peptides
Quantitative protein standards for Western blotting internal controls or ELISA standards.
CHO host cell protein standards are native proteins that can be used as an internal control for the Western blot analysis of CHO Host Cell Proteins or as standards in an ELISA.