Glass Adapters, Ace-Thred to Standard Taper, Ace Glass
Supplier: Ace Glass
5028-226 5028-230 5028-234
76529-624EA 43.34 USD
76529-624 76529-626 76529-628
Glass Adapters, Ace-Thred to Standard Taper, Ace Glass
Adapters Adapters, Conical Ground Joints
Adapter with #7 Ace-Thred and inner joint. Used for probes, thermometers, electrodes, and more.
  • Ground bottom standard taper joint
  • Threaded nylon bushing with O-Ring seal
  • Suitable for vacuum work
  • Nylon bushing, silicone O-Ring
Designed to accept and seal various items such as thermometers, bleed tubes, probes, and electrodes.
Delivery information: Supplied complete with a nylon bushing and silicone O-ring.