Vikan® 59" Aluminum Handle, Transport Line, Remco
Supplier: Remco Products
293752
76528-752EA 42.57 USD
76528-752
Vikan® 59" Aluminum Handle, Transport Line, Remco
Brooms
Professional handle with an insulated sleeve for a comfortable grip.
  • Anodised aluminum handle with black polypropylene grip
  • Euro (hygienic) thread type