Vikan® 51" Aluminum Handle, Remco
Supplier: Remco Products
293566
76528-746EA 30.46 USD
76528-746 76528-748 76528-750
Vikan® 51" Aluminum Handle, Remco
Brooms
Standard broom handle can be used with all brooms, squeegees and scarpers.
  • Aluminum construction with color-coded polypropylene grips
  • Euro (hygienic) thread type
Also suitable for use as a broom handle. It is recommended the handle with attached item should reach the chin of user to prevent back injuries.