HardyDisks™ AST Ofloxacin, 5 µg (OFX-5), Floxin, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z8841
76523-364EA 21.2 USD
76523-364
HardyDisks™ AST Ofloxacin, 5 µg (OFX-5), Floxin, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST), also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.