Abdos® Centrifuge Tubes with Conical Bottom, Sterile, Foxx Life Sciences

Supplier: Foxx Life Sciences
P10416
76521-970CS 229.6 USD
76521-970 76521-972 76521-974 76521-976
Abdos® Centrifuge Tubes with Conical Bottom, Sterile, Foxx Life Sciences
Tubes Centrifuge Tubes
These centrifuge tubes are manufactured using high purity virgin polypropylene (PP) conforming to US FDA 21 CFR free from natural rubber, heavy metal.

  • Cap material: HDPE
  • Silk-screen graduations
  • 15000 RCF max.
  • Color: Natural
  • Temperature range: 0 to 135 °C
  • Autoclavable: Yes
  • Shelf life: 3 years

They are sterilized by gamma radiation.

Certifications: Conforms to US FDA 21 CFR. Complies with: ISO 11137. ISO Standard: 9001, 13485, 14001. Batch certificate included.

Packaging: Paper racked or bulk bag.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR