Evergreen Transport Vial Rack, Caplugs Evergreen
Supplier: Caplugs
240-3300-G10
76521-502CT 61.83 USD
76521-502
Evergreen Transport Vial Rack, Caplugs Evergreen
Racks Chromatography Vial Racks
Caplugs Evergreen Transport Vial Rack is designed to hold up to fifty transport vials of any size.
  • Made up of polystyrene