BioClean™ Non Sterile Tie-on Face Mask, Ansell
Supplier: Ansell Healthcare
MTA210-0
76521-430CS 577.06 USD
76521-430
BioClean™ Non Sterile Tie-on Face Mask, Ansell
Masks Cleanroom Masks
Non sterile tie-on face mask.
  • High bacterial and particle efficiency filtration
  • Fully enclosed malleable nose-band
  • Ultrasonically sealed edges
  • Inner and outer facing layer: non-woven polypropylene/polyethylene (hygroscopic)
Providing high particle and bacterial filtration efficiency, the BioClean™ MTA Cleanroom tie-on face mask is manufactured from cleanroom compatible materials to reduce contamination into the controlled environment. The mask features tie-tapes for a secure fastening.
Certifications: ASTM F2101, ISO 2859, ISO 9001:2015.