BioClean™ Non Sterile DB Pouch-Style Face Mask, Ansell
Supplier: Ansell Healthcare
BDBN
76521-426CS 176.25 USD
76521-426
BioClean™ Non Sterile DB Pouch-Style Face Mask, Ansell
Masks Cleanroom Masks
Non sterile pouch-style face mask.
  • High bacterial and particle filtration efficiency
  • Large breathing chamber
  • Ultrasonically sealed edges
  • Fully enclosed malleable nose-band
  • Inner and outer facing layer: non-woven spunbonded polyester (hygroscopic)
The BioClean™ DB Pouch-style face mask features a large breathing chamber for increased wearer comfort and a malleable noseband for a secure fit. Made from cleanroom compatible materials and ultrasonically sealed edges to reduce the risk of contamination entering the controlled environment.
Certifications: ISO 2859, ISO 9001:2015.