95-Pin Inoculator and Tray, Caplugs
Supplier: Caplugs
222-8600-01A
76521-392CT 238.83 USD
76521-392
95-Pin Inoculator and Tray, Caplugs
Spreaders Inoculating Loops and Needles
Precision molded 95-Pin Inoculator can simultaneously pick up 95 bacterial colony samples (10 µl/pin) and transfer them into or from a 96-well format microplate or microplate tray. The missing pin in the inoculator assembly serves as a blank control.
  • Applications include cell/tissue culture, blood toxicity and serological studies or other dilution and sensitivity tests
  • Each 95-Pin Assembly comes pre-assembled with a 40 ml inoculum tray
  • Additional trays are available separately
  • Both the inoculator assembly and the inoculum tray are gamma sterilized
Ideal for antimicrobial susceptibility testing, urine cultures, bacterial colony subcultures, cell/tissue cultures, cytotoxicity testing, serology dilution studies and evaluation of growth media.