SHOWA CS701 Chemical Resistant Gloves
Supplier: Showa
CS701L-09 CS701M-08 CS701S-07 CS701XL-10 CS701XXL-11
76521-136BG 73.87 USD
76521-136 76521-134 76521-132 76521-138 76521-140
SHOWA CS701 Chemical Resistant Gloves
Gloves
The CS701 food-safe gloves are designed with precision in mind. Anti-slip, rough surface grip gives the wearer ice-grip and optimum performance.
  • Anti-slip grip
  • Chemical-resistant; Oil-resistant
  • Seamless knit
  • Robust durability
  • Forearm protection
  • Natural rubber latex-free
  • Double coated Nitrile
  • Polyester liner; Rough grip
A double-dipped nitrile coating also provides chemical protection and durability to ensure safety from dangerous substances.
The seamless liner and natural latex-free composition are skin-friendly and comfortable for long-wear.
Applications:
- Chemical spray and treatment
- Light chemical handling in horticulture, agriculture
- Coating preparation
- Laboratory, pharma and analysis
- Painting and spray workshops
- Fruit and vegetable processing
- Grain, mill and starch products
- Washing and cleaning
Certifications: CE cat. III, EN 388:2016+A1:2018, EN ISO 374-1:2016+A1:2018/Type A, EN ISO 374-5:2016, EN 407:2004 certified.
Packaging: 10 pair per polybag, 6 polybags per case, 60 pair per case.