VWR® M4 UV/Vis Spectrophotometer
Supplier: VWR International
76520-716
76520-716EA 6136.53 USD
76520-716
VWR® M4 UV/Vis Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The M4 single split-beam spectrophotometer, with flashing xenon lamp, is an easy to use, highly reliable and accurate product.
  • Single split-beam design for sample and blank variability that can be monitored at the same time
  • Xenon flash lamp with a very strong energy in the UV and visible range
  • Connects to a computer based on Windows® through various interfaces
Measure absorbance and transmission at any defined wavelength, generates a graph and searches for peaks and valleys, also analyses the graph via built-in methods.
The application can be extended to luminosity determination, multiple wavelength measurement, quantitative analysis, dynamic determination, DNA/protein determination and spectral measurements.
Delivery information: Supplied with a user manual, 10 mm quartz cuvette, 10 mm glass cuvette, fuse, power cable and dust cover.