VWR® PV4 UV/Visible Spectrophotometer
Supplier: VWR International
76520-714
76520-714EA 3972 USD
76520-714
VWR® PV4 UV/Visible Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The PV4 visible spectrophotometer is an easy to use instrument with advanced performance.
Highly accurate and reliable, with several basic functions, such as photometry test, quantitation test and spectrum scanning.