Evergreen Two Slide Mailers, Caplugs Evergreen
Supplier: Caplugs
240-3072-030
76520-586BG 27.68 USD
76520-586
Evergreen Two Slide Mailers, Caplugs Evergreen
Slide Mailers
Caplugs Evergreen two-slide mailers feature a sturdy living hinge design with friction-fit and post-type locks.
  • Top-panel window allows slides to be viewed without opening
  • Easy-to-use finger/thumb tabs eliminate prying and/or cutting tape
  • Available in multiple colors
A large matte-finished area is molded-in to provide ample room for identification. The slim shape makes it ideal as a component of transport and mailing kits.