Brady®, B33 Series White Polyester with Permanent Acrylic Adhesive Labels, Brady
Supplier: Brady Worldwide
B33-341-422 B33-343-422 B33-1-422 B33-2-422 B33-13-422 B33-15-422 B33-51-422 B33-70-422 B33-3-422 B33-340-422 B33-12-422 B33-37-422 B33-53-422 B33-6-422 B33-7-422 B33-55-422
76520-352RL 164.7 USD
76520-352 76520-354 76520-356 76520-358 76520-360 76520-362 76520-364 76520-366 76520-368 76520-370 76520-372 76520-374 76520-376 76520-378 76520-380 76520-382
Brady®, B33 Series White Polyester with Permanent Acrylic Adhesive Labels, Brady
Labels
Thermal transfer printable, glossy permanent polyester (B-422) labels are ideal for use in all commercial and industrial applications.
  • Recommended ribbon: B30-R6000
  • Heat resistant label with permanent adhesive
  • Permanent acrylic adhesive type
  • Polyester material
  • UL approved, CSA accepted
Aggressive adhesive is good for powder-coated and textured flat surfaces including low surface energy plastics. Designed to withstand exposure to numerous solvents. Service temperature of −40 to 212 °F (−40 to 100 °C). A ribbon is required to print this material. The recommended ribbon is B30-R6000.