Amplitude™ Sigma™ Plus Wipes, Contec
Supplier: Contec
C1E-99
76520-166CS 415.77 USD
76520-166
Amplitude™ Sigma™ Plus Wipes, Contec
Wipes Cleanroom Wipes Dry Cleanroom Wipes
The Amplitude™ Sigma™ Plus wipes are strong and durable, compatible with most solvents, and have outstanding sorbency.
  • Hydroentangled cellulose/poylester provides excellent strength and durability
  • Low levels of particles and extractables
  • Variety of blended fibers enable excellent absorbing capacity
Amplitude™ Sigma™ Plus Wipes are constructed using a nonwoven fabric that is a hydroentangled blend of polyester and cellulose fibers. Wet or dry, the wipe maintains a high level of strength and durability. Specially treated to reduce particles and fibers, it is recommended when low particle generation combined with sorbency is of high priority.