VWR® USB Digital Data Logger (DDL) (Dual Probe)
Supplier: VWR International
76520-154
76520-154EA 335.19 USD
76520-154
VWR® USB Digital Data Logger (DDL) (Dual Probe)
Data Loggers
This USB data logger is compliant with current CDC vaccine storage monitor guidelines. It includes a buffered probe in product simulated solution, °F/°C switchable readout, a detachable dual probe, audible and visual alarms,and temperature data downloadable via USB drive (not included). This device stores up to 3 years of data.
  • Material: Assorted materials