VWR® USB Digital Data Logger (DDL) (Single Probe)
Supplier: VWR International
CP-DDL1-18
76520-152EA 311.78 USD
76520-152
VWR® USB Digital Data Logger (DDL) (Single Probe)
Data Loggers
This USB data logger is compliant with current CDC vaccine storage monitor guidelines. It has a buffered probe in product simulated solution, °F/°C switchable readout, a detachable single probe, audible and visual alarms, and temperature data downloadable via USB drive (not included). Device stores up to 3 years of data.
  • Material: Assorted materials