POLYPROBE DRY WHIRL-PAK BX100
Supplier: NASCO
PolyProbe Dry, Whirlpack, esigned for surface sampling and effective in hard-to-reach areas, the 8 in. Inch (20.3 cm) long probe consists of a polypropylene holder with detachable handle that is adhered to a durable polyurethane sponge that resists tearing and fraying
Learn more
About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the is still displayed and you need assistance, please call us at 1-800-932-5000.
To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- State issued document with your organization's Federal Tax ID Number
- State issued document with your organization's Resale Tax ID Number
- City or County issued Business License
- State Department of Health Services License
- Any other ID issued by the State that includes the business name & address