Muffle Furnaces with Fold Down Door, Tmax 1100 °C and Controller R7, LE Series (LE 1/11 - LE 24/11), Nabertherm
Supplier: NABERTHERM INC.
LE241K17N1-240
76516-384EA 7719.7 USD
76516-384
Muffle Furnaces with Fold Down Door, Tmax 1100 °C and Controller R7, LE Series (LE 1/11 - LE 24/11), Nabertherm
Ovens Furnaces (Ashing and Muffle Ovens)
With their convincing price/performance ratio and the fast heat-up rates, these compact muffle furnaces are perfect for many applications in the laboratory. Quality features like the dual shell furnace housing of rust-free stainless steel, their compact, lightweight constructions, or the heating elements encased in quartz glass tubes make these models reliable partners for your application.
  • Heating from two sides from heating elements protected in quartz glass tubes
  • Fast heating times, housing coated in RAL
  • Maintenance-friendly replacement of heating elements and insulation
  • Solid state relays provide for lownoise operation
  • Flap door which can also be used as a work platform
  • Exhaust air outlet in rear wall
  • Defined application within the constraints of the operating instructions
Compact dimensions and light weight. Controller mounted under the door to save space, controller R7. The furnaces have Tmax 1100 °C.
Dual shell housing made of textured stainless steel sheets with additional fan cooling for low surface temperature. Exclusive use of insulation materials without categorization according to EC Regulation No. 1272/2008 (CLP).