Soft Care® Impact Foam Instant Hand Sanitizer, Diversey
Supplier: DIVERSEY INC.
101104202
76514-360CS 325.06 USD
76514-360
Soft Care® Impact Foam Instant Hand Sanitizer, Diversey
Skincare Soaps and Skin Cleansers
Skin softening emollients leave hands clean and soft with no sticky residue.
  • Contains 76% (v/v), 72% (w/w) ethyl alcohol, meeting APIC, PHAC, CDC and WHO healthcare personal hand wash guidelines
  • Made with 96% biobased ingredients
  • Dye-free and fragrance-free formulas reduce potential risk of skin irritation and allergic reactions for users with sensitive skin
  • NSF E-3 certified for use in food handling establishments
Soft Care® impact is an alcohol-based instant hand sanitizer enriched with skin conditioning emollients to keep skin soft and protect against irritation for even the most frequent users. Fragrance-free and dye-free, Soft Care® impact meets CDC, WHO, APIC and PHAC hand hygiene recommendations for healthcare personnel hand rubs.