Corepoint Scientific® White Diamond Series Undercounter Laboratory Freezers, Horizon Scientific
Supplier: Horizon Scientific
PF051WWW/0M20
76514-186EA 1659.13 USD
76514-186
Corepoint Scientific® White Diamond Series Undercounter Laboratory Freezers, Horizon Scientific
Freezers Laboratory / General Purpose Freezers
These White Diamond undercounter freezers are designed for a variety of laboratory purposes.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port
  • Material: White cabinet and door
  • Natural, hydrocarbon refrigerant
These units utilize state-of-the-art technology including microprocessor temperature controllers, digital temperature displays, interior lighting, and a series of temperature alarms. Ideal for laboratory use.
All units include a three year parts and labor warranty, plus an additional two year compressor parts warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency.
Certifications: UL/C-UL listed.