Corepoint Scientific® General Purpose Refrigerator and Freezer Combo Units, Horizon Scientific
Supplier: Horizon Scientific
LRF151WWW/0HC
76514-178EA 2142.64 USD
76514-178
Corepoint Scientific® General Purpose Refrigerator and Freezer Combo Units, Horizon Scientific
Refrigerators
These General Purpose Refrigerator and Freezer Combination models are designed for laboratory use.
  • 14 cu. ft. capacity
  • ADA compliant
  • Auto defrost freezer
  • Two probe access ports
  • Material: White cabinet and door
  • Natural, hydrocarbon refrigerant
These combo units offer a cold-storage solution for a variety of laboratory needs. The units utilize mechanical thermostats, adjustable storage bins and shelving, LED interior lighting and probe access ports for both the refrigerator and freezer. Ideal for laboratory use.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty. They run on natural, hydrocarbon refrigerant for environmental health and economic efficiency.