Orum International TRIO.BAS™ Satellite with Tri-Clamp Connection, Contact Plate Format
Supplier: Hardy Diagnostics
BAS320K
76514-146EA 3493.67 USD
76514-146
Orum International TRIO.BAS™ Satellite with Tri-Clamp Connection, Contact Plate Format
Samplers Air Samplers
A customizable air sampling satellite option for use with RABS ISOLATOR or MULTIFLEX instruments.
  • Ideal for use in cleanroom barrier systems where space is limited
Note: Third image displays the Tri-Clamp Satellite shown in a horizontal position with stainless steel AISI 316 piping connection and MULTIFLEX 1 instrument (both sold separately).