Anti-Sarilumab Mouse Monoclonal Antibody [clone: 23H12]

Supplier: Genscript
A02043-40
76510-106EA 153.9 USD
76510-106
Anti-Sarilumab Mouse Monoclonal Antibody [clone: 23H12]
Antibodies
The product is specific for sarilumab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A02042-40, anti-sarilumab antibody (26B10), mAb, mouse.

  • Anti idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type: Primary
Antigen: Sarilumab
Clonality: Monoclonal
Clone: 23H12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR