Anti-Infliximab Mouse Monoclonal Antibody [clone: 23H5]

Supplier: Genscript
A02041-40
76510-102EA 153.9 USD
76510-102
Anti-Infliximab Mouse Monoclonal Antibody [clone: 23H5]
Antibodies
The product is specific for infliximab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A02040-40, anti-infliximab antibody (27A4), mAb, mouse.

  • Anti idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
.

Type: Primary
Antigen: Infliximab
Clonality: Monoclonal
Clone: 23H5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR