Anti-Nivolumab Rabbit Monoclonal Antibody (Biotin) [clone: 40F11]

Supplier: Genscript
A01989-40
76510-002EA 219.4 USD
76510-002
Anti-Nivolumab Rabbit Monoclonal Antibody (Biotin) [clone: 40F11]
Antibodies
The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A01987-40, MonoRab anti-nivolumab antibody (33H4), mAb, rabbit.

  • Anti idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
.

Type: Primary
Antigen: Nivolumab
Clonality: Monoclonal
Clone: 40F11
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR