Anti-Tocilizumab Mouse Monoclonal Antibody [clone: 6C10]

Supplier: Genscript
A01979-40
76509-984EA 153.9 USD
76509-984
Anti-Tocilizumab Mouse Monoclonal Antibody [clone: 6C10]
Antibodies
The product is specific for tocilizumab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A01978-40, anti-tocilizumab antibody (14B10), mAb, mouse.

  • Anti idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.
.

Type: Primary
Antigen: Tocilizumab
Clonality: Monoclonal
Clone: 6C10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR