Anti-Bevacizumab Mouse Monoclonal Antibody [clone: 6C3]

Supplier: Genscript
A01976-40
76509-978EA 153.9 USD
76509-978
Anti-Bevacizumab Mouse Monoclonal Antibody [clone: 6C3]
Antibodies
The product is specific for Bevacizumab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A01975-40, Anti-Bevacizumab Antibody (4H1), mAb, Mouse.

  • Anti-Idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type: Primary
Antigen: Bevacizumab
Clonality: Monoclonal
Clone: 6C3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR