Anti-Nivolumab Mouse Monoclonal Antibody [clone: 6G5]

Supplier: Genscript
A01965-40
76509-958EA 153.9 USD
76509-958
Anti-Nivolumab Mouse Monoclonal Antibody [clone: 6G5]
Antibodies
The product is specific for Nivolumab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A01847-40, Anti- Nivolumab Antibody (8G6G3D8), mAb, Mouse.

  • Anti-Idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type: Primary
Antigen: Nivolumab
Clonality: Monoclonal
Clone: 6G5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR