Anti-Atezolizumab Mouse Monoclonal Antibody [clone: 6B12] (Biotin)

Supplier: Genscript
A01950-40
76509-928EA 153.9 USD
76509-928
Anti-Atezolizumab Mouse Monoclonal Antibody [clone: 6B12] (Biotin)
Antibodies
The product is specific for Atezolizumab. The antibody is recommended as a detection antibody in a pharmacokinetic (PK) bridging assay with capture antibody GenScript, A01948-40, Anti-Atezolizumab Antibody (10G9), mAb, Mouse.

  • Anti-Idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type: Primary
Antigen: Atezolizumab
Clonality: Monoclonal
Clone: 6B12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR