Anti-Rituximab Rabbit Monoclonal Antibody [clone: 137C6]

Supplier: Genscript
A01942-40
76509-912EA 219.4 USD
76509-912
Anti-Rituximab Rabbit Monoclonal Antibody [clone: 137C6]
Antibodies
The product is specific for Rituximab. The antibody is recommended as a capture antibody in a pharmacokinetic (PK) bridging assay with detection antibody GenScript, A01944-40, MonoRab™ Anti-Rituximab Antibody (194D7) [Biotin], mAb, Rabbit.

  • Anti-Idiotype antibodies
  • High specificit
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type: Primary
Antigen: Rituximab
Clonality: Monoclonal
Clone: 137C6
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR