Anti-IgG Mouse Monoclonal Antibody [clone: 1H7A8]

Supplier: Genscript
A01011
76509-810EA 85.95 USD
76509-810
Anti-IgG Mouse Monoclonal Antibody [clone: 1H7A8]
Antibodies
Mouse IgG1 control (whole molecule), purified is suitable to be used as control, standard, blocking agent, or coating protein in a variety of assays, including ELISA, FACS, ICC immunoblotting, dot blot, western blot, immunoprecipitation, immunodiffusion, and immunoelectrophoresis. It may also be used as antigen or ligand in immunochemical conjugation reaction. GenScript Mouse IgG1 control (whole molecule), purified is highly purified from culture supernatant from roller bottle through protein A affinity column.GenScript mouse IgG1 control (whole molecule), purified is produced from the hybridoma resulting from fusion of SP2/0-Ag14 myeloma and B-lymphocytes.

  • High specificit
  • High affinity
  • High stability

GenScript offers a wide selection of polyclonal and monoclonal loading control antibodies for α-tubulin and β-actin for several species. HRP- and biotin-conjugated versions of these antibodies are also available. GenScript also offers a series of isotype control antibodies that can be used in your research.
.

Type: Control 
Antigen: IgG
Clonality: Monoclonal
Clone: 1H7A8
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR