NEBNext® ARTIC SARS-CoV-2 RT-PCR Module, New England Biolabs
Supplier: New England Biolabs (NEB)
E7626S E7626L
76509-468EA 80 USD
76509-468 76509-470
NEBNext® ARTIC SARS-CoV-2 RT-PCR Module, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The ARTIC SARS-CoV-2 protocol from the ARTIC Network is a multiplexed amplicon approach covering the whole viral genome.
  • Updated primers for effective Omicron coverage
  • Effective with a wide range of viral genome inputs (10 to 10000 copies)
  • Express protocol option, with fewer cleanups
The NEBNext ARTIC SARS-CoV-2 RT-PCR Module is based on this method and incorporates optimized reagents for the cDNA synthesis and targeted cDNA amplification steps. A single RT protocol is used regardless of input amount (10 to 10000 viral genome copies), and no normalization step is required ahead of targeted amplification. The multiplex primers are balanced to provide uniform amplicon coverage across the viral genome.