96 Square Well EVA Sealing Mat, Irish Life Sciences
Supplier: Irish Life Sciences
SM428
76509-444CS 280.23 USD
76509-444
96 Square Well EVA Sealing Mat, Irish Life Sciences
Seals Microplate Seals
Sealing Mat suitable for sealing 96 Square Well plate. Made from EVA. Ideal for retaining the sample in the well during transit or to stop the well and its contents from becoming contaminated.
  • Suitable for long term storage
  • High chemical resistance
  • Free from RNase, Dnase
Mats are an economic alternative to thermal or adhesive seals, constructed of high quality ethylene-vinyl acetate (EVA).
Certifications: Manufactured in ISO Class 8 Cleanroom.