MD8 AirPort Portable Air Sampler, Sartorius
Supplier: Sartorius
16757
76509-354EA 5541.63 USD
76509-354
MD8 AirPort Portable Air Sampler, Sartorius
Samplers Air Samplers
MD8 Airport portable, battery operated air monitoring system for the quantitative and reproduceable collection of airborne bacteria, yeasts, and viruses. For use with gelatine filter disposables or BactAir media plates.
  • Battery power level clearly indicated
  • Ergonomically designed and easy to clean
  • Utilizes Gelatin Membrane Filter Method, for reliable and accurate results
  • MD8 Airport can also be used with BACTair culture media plates
  • User friendly prompting with the option of four languages (English, French, German, and Spanish)
  • Flexible options for customized use including adjustable air throughput and air flow rate
  • Parameters last used are stored even after automatic shut-off
  • Can be calibrated on-site
A fundamental aspect of cleanrooms and critical zones is to achieve the lowest concentrations of airborne particles by reducing ther introduction and generation. Measurements for this purpose must be accurate as well as reliable, and must not interfere with the zone protection.
The portable MD8 Airport air sampler is designed to detect the smallest viruses and microorganism in the air by using the unique Gelatine Membrane Filters (GMF) and BACTair agar plates.
Certifications: CSA compliant.
Ordering information: Accessories not included. Adapters must be ordered separately.
Delivery information: Delivered with battery charger and country specific plugs.