Microsart® e.jet Pump
Supplier: Sartorius
166MP4
76509-334EA 2114.61 USD
76509-334
Microsart® e.jet Pump
Pumps Vacuum Pumps
The Microsart® e.jet is a laboratory vacuum pump which creates vacuum and concomitantly transfers the filtered liquid to waste.
  • For vacuum filtration in microbiological analysis
Microsart® e.jet is ideal for sample preparation in microbiological analysis achieving a trans membrane pressure of 600 mbar and a flow rate of 4.0 Nl/min (4.0 Normlitre water displacement by air in 1 minute). Constant flow rates and a defined maximum vacuum guarantees smooth and reliable filtration.