Orum International Cleanroom Bag for Sterile Transport of Media Plates
Supplier: Hardy Diagnostics
BAS381
76509-296PK 465.95 USD
76509-296
Orum International Cleanroom Bag for Sterile Transport of Media Plates
Bags Storage Bags
Sterile bags for transporting petri dishes, contact plates, swabs, or other objects outside of the cleanroom.
  • Sterility traceability
  • Triple-wrapped and individually sealed to ensure sterility
  • Beta sterilization certificate included
  • Color is easy to identify in the cleanroom
  • Write-on oversized area to identify sample contents suitable for use with permanent markers
Self-adhesive flap closure prevents contamination after samples are placed inside of the bag. Bags feature an easy-open tear line which guarantees the sterility of the single bag. The elongated, flat bag design facilitates stacking of multiple bags, easy reading of bag contents, and allows plate lids to stay secured inside the bag during handling.