Orum International TRIO.SETTLE Table-top Holder for Petri Settling Plate, Hardy Diagnostics
Supplier: Hardy Diagnostics
BAS367
76509-292EA 1624.83 USD
76509-292
Orum International TRIO.SETTLE Table-top Holder for Petri Settling Plate, Hardy Diagnostics
Racks Petri Dish Racks
Benchtop format culture media plate holder with an upper surface to hold open media plate during 'passive' sampling, and a lower surface area to accommodate the lid of the media plate.
  • Constructed from AISI 316 stainless steel
  • Can be angled to direct the media plate’s exposure to unidirectional airflow