LabRobot Dilucup™ Elegance Butterfield's Phosphate Buffer
Supplier: HARDY DIAGNOSTICS
40DEBFD6
76509-256EA 548.93 USD
76509-256 76509-258
LabRobot Dilucup™ Elegance Butterfield's Phosphate Buffer
Buffers Biological Buffers
Butterfield's Phosphate Buffer is used as a diluent for the preparation of dilutions in plate count and other laboratory procedures.
  • Sterile, pre-filled diluent
  • Easy peel-off film
  • Long shelf life
A Dilucup is a small container pre-filled with 9 ml of a sterile diluent. This system maximizes lab efficiency for serial dilutions, with its pre-filled, sterile, and ready-to-use dilution cups.
For industries that perform serial dilutions, the Dilucup Elegance serial dilution system is the easiest, most cost-effective answer in the field. These save time and standardize results. You will also see measurable differences in the amount of storage space required for this product line. The compact and thoughtfully design packaging will save you on shipping and shelf space. The Dilucup line also reduces the occurrence of repetitive motion injuries.