Safety Funnels with Lid 'MARCO' V2.0, Electrically Conductive, SCAT Europe
Supplier: SCAT AMERICAS, INC.
306980 317637 317651 318630 318960 318961 318962 318964 318980 318981 318983 318984 318985
76503-740EA 172.88 USD
76503-740 76503-880 76503-902 76503-904 76503-918 76503-920 76503-922 76503-924 76503-926 76503-928 76503-930 76503-932 76503-934
Safety Funnels with Lid 'MARCO' V2.0, Electrically Conductive, SCAT Europe
Closures Safety Caps and Safety Waste Caps
S.C.A.T. 'MARCO' V2.0 safety funnels can be closed safely and prevent danger of ignition.
  • 'MARCO', V2.0 safety lid funnel is a modern lid funnel made of high-quality HDPE with extremely high resistance to acids, alkalis and many chemicals
  • Integrated gripping edge on the funnel lid increases the grip without having to touch the possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes the funnel edge and prevents explosive or health-endangering gases escaping
  • Sturdy hinge guarantees the closing mechanism has a long service life
  • Rotating screw cap makes it easier to screw the lid funnel on and off the canister
  • With removable sieve for easy cleaning
  • The non-conductive version of the 'MARCO' V2.0 safety lid funnel is in line with the SCAT color scheme with its blue and white coloring, the conductive version is black, this difference in color helps differentiate the conductive from the non-conductive version
The innovative 'MARCO', V2.0 safety funnel is a high-quality lid funnel for the disposal of liquid waste. The lid, with an integrated rubber lip, seals the funnel and prevents the escape of toxic or explosive vapors. Numerous features guarantee safe handling. These electrically conductive versions has an earth connection to avoid ignition hazards. Available in different thread sizes for safe attachment to laboratory bottles, canisters or other containers.
Diameter: 140 mm
Delivery information: Supplied with grounding cable.