Funnel with Ball and Lid, ARNOLD, V2.0, SCAT
Supplier: SCAT AMERICAS, INC.
317632 317641 317642 317643 317644 317645 317646 317647 317648 317649
76503-874EA 293.91 USD
76503-874 76503-884 76503-886 76503-888 76503-890 76503-892 76503-894 76503-896 76503-898 76503-900
Funnel with Ball and Lid, ARNOLD, V2.0, SCAT
Funnels Safety Funnels
The strong safety funnel 'ARNOLD' V2.0 with lid and ball meets the highest requirements for safe disposal of hazardous liquid waste due to its combination of two independent safety mechanisms. The two safety features consist of a ball integrated in the funnel basin and a sealing ring incorporated in the hinged lid of the funnel, which prevent the escape of toxic or explosive gases. The new design ensures greater stability, durability and prevents deformation of the funnel.
  • Made of high quality white/blue HDPE material for maximum stability and durability
  • Hinged lid with sealing ring for closure when the funnel is not in use
  • Optimized sieve for catching larger particles and for depositing laboratory bottles
  • Improved splash guard with integrated air channels to prevent bubble formation
  • Ball valve with automatic opening mechanism during filling
  • Individually shortenable safety lance for clean drainage of liquid waste
  • All essential components are removable for easy cleaning
The safety funnel 'ARNOLD' V2.0 with lid and ball valve is available for different container threads. In order to be able to produce in an environmentally friendly and economical way, we supply individual variants with a thread adapter, which has no influence on the functionality or safety of the funnel.
Delivery information: Including lance (220 mm), splash guard and removable sieve.