Level Control Systems with Signal Boxes, SCAT
Supplier: SCAT AMERICAS, INC.
108087 108088 108119 108121 108122 108125 108157 108158 108159
76503-480EA 687.37 USD
76503-480 76503-482 76503-492 76503-494 76503-496 76503-500 76503-528 76503-530 76503-532
Level Control Systems with Signal Boxes, SCAT
Level Sensors
Monitor two filling levels at once, perfect integration into lab furniture with the Built-In Signalbox ant the Signalbox T1 and T4.
  • For all types of Safety Waste Caps and other electronic level controllers
  • Touchless alarm-resetting fields for two containers
  • 2 switch inputs and 2 switch outputs for peripheral units
LEDs and acoustic buzzer warn you in due time before the filling level of your collection containers reaches a critical height. When the container is changed, the alert can be muted via the new touchpad. Pumps and other external devices can be conveniently triggered via the integrated interface.