Lab Installations for SCAT SafetyCaps, S.C.A.T.
Supplier: SCAT AMERICAS, INC.
106406 106502 106677 107836 107886 108124 108278 160145 307508
76502-868EA 32.13 USD
76502-868 76502-938 76503-010 76503-300 76503-312 76503-498 76503-574 76503-682 76503-798
Lab Installations for SCAT SafetyCaps, S.C.A.T.
Chromatography Supplies Chromatography Spare Parts
SCAT lab installations can be completely integrated into lab furniture and offer a gastight system for safe fluid management.
SCAT lab installations fit any SCAT SafetyCap or SafetyWasteCap with GL 45 thread size, to effectively prevent health hazards and environmental damage. Changeable exhaust filter and tube connectors can be fixed directly onto the desktop for quick and easy access. The waste container can be stored under the desk, in the cupboard or sideways.
Built-in air valves and exhaust filters shut away hazardous vapors and gases, even during supply and disposal of solvents and waste fluids.
Most components are made of electroconductive HDPE and can be grounded safely.
More assembly options are available. Contact your Avantor representative for more information.