SymLine® Built-in Signalbox and Switch Amplifiers, S.C.A.T.
Supplier: SCAT AMERICAS, INC.
106590 108219
76502-986EA 260.3 USD
76502-986 76503-560
SymLine® Built-in Signalbox and Switch Amplifiers, S.C.A.T.
Level Sensors
Monitor two filling levels at once. Perfect integration into lab furniture with the Built-In signalbox and switch amplifier.
  • Monitor two filling levels at once
  • Touchpad control
  • Level control through status LEDs
  • Optical and acoustic alarm signals
  • Compatible with switch amplifier
LEDs and acoustic buzzer provide warning in due time before the filling level of collection containers reaches a critical height. When the container is changed, the alert can be muted via the new touchpad. Pumps and other external devices can be conveniently triggered via the integrated interface. According to ATEX Directives, Ex-sensors have to be installed in conjunction with a switch amplifier when used in explosion-prone areas (Ex-zones). Signalboxes and switch amplifier must be placed outside of the Ex-zone.