SymLine® Grounding and Adapters
Supplier: SCAT AMERICAS, INC.
106408 106409 106410 106415 106417 106423 106434 106510 106523 106539 106584 107632 108079 108176 108262 108270 108272 121211
76502-872EA 32.13 USD
76502-872 76502-874 76502-876 76502-880 76502-882 76502-886 76502-894 76502-942 76502-946 76502-950 76502-984 76503-222 76503-476 76503-540 76503-566 76503-568 76503-570 76503-652
SymLine® Grounding and Adapters
Fittings and Connectors
Dovetail the SymLine® system to suit specific requirements. This product selection includes several useful ancillaries.
  • SymLine® offers a suitable adapter for every common thread, to eliminate problems connecting different thread types
  • Many useful accessories allow users to configure the SymLine® system according to specific needs
  • All accessories are eclectically dissipative and approved for use in laboratories with explosion hazards
Dispose of liquid HPLC waste safely. SymLine® is quickly installed without drilling holes or making technical changes to existing laboratory equipment.