SymLine® Safety Waste Caps and Ventilation Kit, SCAT
Supplier: SCAT AMERICAS, INC.
106478 106480 106484 106507 106522 106527 306482 306494 306498 306509 306581
76502-928EA 428.11 USD
76502-928 76502-930 76502-932 76502-940 76502-944 76502-948 76503-730 76503-732 76503-734 76503-736 76503-738
SymLine® Safety Waste Caps and Ventilation Kit, SCAT
Closures Safety Caps and Safety Waste Caps
Safety waste caps safely connect pipework to the waste container. The integrated exhaust filter blocks solvent vapors, for the protection of health and the environment.
  • Caps are available with different thread sizes for attachment to the pipe system
  • Caps are available with different numbers of hose connections for disposal of liquid waste
  • By using a 4-in-1 collector, up to 4 safety waste caps can be connected simultaneously to only one connection point on the pipe system
  • Ventilation tube, length 1500 mm, allows connection to lab exhaust system
  • Adapter (Y-connector) for connecting two ventilation tubes to one common exhaust tube
Optionally, safety waste caps are available with electronic level sensors that indicate the degree of filling of the disposal container.