Five-Slide Mailer, Caplugs
Supplier: Caplugs
240-3074-030
76502-820CT 31.75 USD
76502-820
Five-Slide Mailer, Caplugs
Slide Mailers
The Five-Slide Mailer features an integral recessed lid design which limits slide movement during transit to prevent accidental breakage.
  • Rounded corners allow easier handling
  • Eyelet is provided for optional security ties and/or identification tags
  • Mailers can also function as miniature Coplin jars
Lightweight design (less than ¹/₃ oz.) helps to minimize transport costs.